tri-Calcium phosphate (35 - 40% Ca) for analysis
Supplier: Thermo Scientific Chemicals
AC21348-0010
AC21348-0050
TS21348-0010EA
128
USD
TS21348-0010
TS21348-0050
tri-Calcium phosphate (35 - 40% Ca) for analysis
tri-Calcium phosphate
Formula:
Ca₃(PO₄)₂ Melting Pt: 1670 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015984 CAS Number: 7758-87-4 EINECS: 231-840-8 Merck Index: 12,01741 |
Specification Test Results
Appearance | White Powder |
Titration Complexometric | 35 to 40 % (Ca) |
Chloride (Cl) | ≤0.14 % |
Fluoride (F) | ≤0.0075 % |
Carbonate (CO3) | Passes test |
Nitrate (NO3) | Passes test |
Sulfate (SO4) | ≤0.5 % |
Arsenic (As) | ≤0.0003 % |
Barium (Ba) | Passes test |
Iron (Fe) | ≤0.04 % |
Heavy metals (as Pb) | ≤50 ppm |
Insoluble matter | ≤0.2 % (in HCl) |
Loss on ignition | ≤8.0 % (800°C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...