Nickel(II) bromide 99%

Supplier: Thermo Scientific Chemicals
Danger

AC20336-0100 AC20336-0500 AC20336-2500
TS20336-0100EA 41 USD
TS20336-0100 TS20336-0500 TS20336-2500
Nickel(II) bromide 99%
Nickel(II) bromide
Formula: Br₂Ni
Melting Pt: 963 °C
Storage Temperature: Ambient
MDL Number: MFCD00011141
CAS Number: 13462-88-9
EINECS: 236-665-0
UN: 3077
ADR: 9,III
Merck Index: 12,06585

Order Now

Specification Test Results

Appearance Brown Crystalline powder and/or chunks
Titration Argentometric ≥98.5 % (on dry substance)
Water ≤6 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR