Calcium carbonate 99%, extra pure
Supplier: Thermo Scientific Chemicals
AC45068-0010
AC45068-0050
AC45068-0250
TS45068-0010EA
27
USD
TS45068-0010
TS45068-0050
TS45068-0250
Calcium carbonate 99%, extra pure
Calcium carbonate
Formula:
CaCO₃ Melting Pt: 825 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010906 CAS Number: 471-34-1 |
Specification Test Results
Appearance | White to almost white Powder |
Titration Complexometric | 98.5 to 101.5 % |
Loss on drying | ≤2.0 % (1 g, 200°C) |
Insoluble matter | ≤0.2 % (in dilute acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...