Nickel acetate tetrahydrate 99%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nickel diacetate tetrahydrate, Nickel (II) acetate tetrahydrate, Nickel di(acetate) tetrahydrate, Acetic acid nickel (II) salt tetrahydrate
AC41554-0050
AC41554-1000
AC41554-5000
TS41554-0050EA
33
USD
TS41554-0050
TS41554-1000
TS41554-5000
Nickel acetate tetrahydrate 99%, Extra Pure
Nickel acetate tetrahydrate
Formula:
C₄H₆NiO₄·4H₂O MW: 248.84 g/mol Melting Pt: 250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066973 CAS Number: 6018-89-9 EINECS: 206-761-7 UN: 3077 ADR: 9,III Merck Index: 12,06583 |
Specification Test Results
Appearance | Green to blue Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥98.5 % |
Cobalt (Co) | ≤0.2 % |
Iron (Fe) | ≤20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...