Lead(II) sulphide 90%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead sulphide

AC45397-0010 AC45397-0050
TS45397-0010EA 96 USD
TS45397-0010 TS45397-0050
Lead(II) sulphide 90%, pure
Lead(II) sulphide
Formula: ̍PbS
Boiling Pt: 1281.1 °C
Melting Pt: 1113.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00016280
CAS Number: 1314-87-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Gray to purple Powder
Assay ≥78 % (Pb)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR