Ammonium fluoride 98+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Fluoroammonium

AC20184-0025 AC20184-1000 AC20184-5000
TS20184-0025EA 282 USD
TS20184-0025 TS20184-1000 TS20184-5000
Ammonium fluoride 98+%, Extra Pure
Ammonium fluoride
Formula: NH₄F
MW: 37.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011423
CAS Number: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1,III
Merck Index: 13,00524

Order Now

Specification Test Results

Appearance White Adhering crystalline powder
Residue after ignition ≤0.01 % (SO4)
Titration excess NaOH ≥98.0 %
Hexafluorosilicate (SiF6) ≤0.1 %
Heavy metals (as Pb) ≤5 ppm
(NH4)HF2 ≤1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR