Ammonium fluoride 98+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Fluoroammonium

AC42329-0025 AC42329-0050 AC42329-5000
TS42329-0025EA 329 USD
TS42329-0025 TS42329-0050 TS42329-5000
Ammonium fluoride 98+% ACS
Ammonium fluoride
Formula: NH₄F
MW: 37.04 g/mol
Melting Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00011423
CAS Number: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1,III
Merck Index: 13,00524

Order Now

Specification Test Results

Appearance White Crystalline powder
Residue after ignition ≤0.01 %
Titration excess NaOH ≥98.0 %
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR