Copper(II) chloride dihydrate 99%, powder, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate

AC40584-0025 AC40584-0050 AC40584-5000
TS40584-0025EA 260 USD
TS40584-0025 TS40584-0050 TS40584-5000
Copper(II) chloride dihydrate 99%, powder, extra pure
Copper(II) chloride dihydrate
Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Melting Pt: 100 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660

Order Now

Specification Test Results

Appearance Blue-green to blue Crystalline powder or needles
Titration Iodometric ≥98.5 %
Nitrate (NO3) ≤200 ppm
Sulfate (SO4) ≤1000 ppm
Iron (Fe) ≤300 ppm
Zinc (Zn) ≤500 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR