Tin(II) chloride, anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC19698-0025
AC19698-0250
AC19698-1000
AC19698-5000
TS19698-0025EA
677
USD
TS19698-0025
TS19698-0250
TS19698-1000
TS19698-5000
Tin(II) chloride, anhydrous 98%
Tin(II) chloride
Formula:
SnCl₂ Boiling Pt: 623 °C (1013 hPa) Melting Pt: 246 °C Density: 3.95 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011241 CAS Number: 7772-99-8 UN: 3260 ADR: 8,III |
Specification Test Results
Appearance | White to off-white Crystalline powder or flakes |
Titration Iodimetric | 97.5 to 102.5 % |
Iron (Fe) | ≤50 ppm |
Lead (Pb) | ≤75 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...