Sodium tetrachloropalladate(II) (∼36% Pd)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Palladium sodium chloride, Sodium palladium (II) chloride, Disodium tetrachloropalladate
AC19513-0010
AC19513-0050
AC19513-0250
TS19513-0010EA
119
USD
TS19513-0010
TS19513-0050
TS19513-0250
Sodium tetrachloropalladate(II) (∼36% Pd)
Sodium tetrachloropalladate(II)
Formula:
Cl₄Na₂Pd MW: 294.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003487 CAS Number: 13820-53-6 EINECS: 237-502-6 UN: 1759 ADR: 8,III |
Specification Test Results
Appearance | Green-brown to brown Powder or granules and/or chunks |
Assay | 35 to 37 % (Pd) |
Total trace metal impurities | ≤300 ppm (Pt + Ru + Rh + Ir) |
Calcium (Ca) | ≤50 ppm |
Copper (Cu) | ≤50 ppm |
Gold (Au) | ≤100 ppm |
Iron (Fe) | ≤50 ppm |
Magnesium (Mg) | ≤50 ppm |
Silicon (Si) | ≤50 ppm |
Silver (Ag) | ≤100 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...