Lithium nitrate 99.999% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC21254-0050
AC21254-0250
AC21254-1000
TS21254-0050EA
50
USD
TS21254-0050
TS21254-0250
TS21254-1000
Lithium nitrate 99.999% (trace metals basis)
Lithium nitrate
Formula:
LiNO₃ MW: 68.95 g/mol Boiling Pt: 600 °C (760 mmHg) Melting Pt: 251 °C Density: 2.38 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011094 CAS Number: 7790-69-4 UN: 2722 ADR: 5.1,III |
Specification Test Results
Appearance | White Crystalline powder and/or chunks |
Total trace metal impurities | ≤10 ppm |
Lithium (Li) | 9 to 11 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...