Copper(II) pyrithione

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Copper Omadine, Bis(1-Hydroxy-1H-pyridine-2-thionato-O,S)copper, Bis(2-pyridinethiol 1-oxide)copper

AC45721-0010 AC45721-0050
TS45721-0010EA 101 USD
TS45721-0010 TS45721-0050
Copper(II) pyrithione
Copper(II) pyrithione
Formula: C₁₀H₈CuN₂O₂S₂
MW: 315.85 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02660770
CAS Number: 14915-37-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Green Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR