Chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Chlorodihydrido[bis(2-di-i-propylphosphinoethyl)amine]iridium(III), CHLORODIHYDRIDO[BIS(2-DI-I-PROPYLPHOSPHINOETHYL)AMINE]IRIDIUM (III), CHLORODIHYDRIDO[BIS(2-DI-I-PROPYLPHOSPHINOETHYL)AMINE]IRIDIUM(III)

AC43512-0010 AC43512-2500
TS43512-0010EA 322 USD
TS43512-0010 TS43512-2500
Chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium 97%
Chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium
Formula: C₁₆H₄₂ClIrN₂P₂
Melting Pt: 240.6…241.9 °C
Storage Temperature: Ambient
MDL Number: MFCD07782018
CAS Number: 791629-96-4

Order Now

Specification Test Results

Appearance White to tan Powder
Infrared spectrum Conforms
Elemental Analysis 2.50 to 2.70 % (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR