Tris(trimethylsilyl)amine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nonamethyltrisilazane, Silanamine,1,1,1-trimethyl-N,N-bis(trimethylsilyl)-, Amine, tris(trimethylsilyl)-, Nitrilotris(trimethylsilane)

AC37877-0050 AC37877-0250
TS37877-0050EA 46 USD
TS37877-0050 TS37877-0250
Tris(trimethylsilyl)amine 99%
Tris(trimethylsilyl)amine
Formula: C₉H₂₇NSi₃
Boiling Pt: 72 °C (10 mmHg)
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00047990
CAS Number: 1586-73-8
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless Crystals
Infrared spectrum Conforms
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR