Vinylmagnesium chloride 16,5% (w/w) 1.9 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Magnesium,chloroethenyl-

AC25259-1000 AC25259-8000
TS25259-1000EA 90 USD
TS25259-1000 TS25259-8000
Vinylmagnesium chloride 16,5% (w/w) 1.9 M in THF, AcroSeal™
Vinylmagnesium chloride
Formula: C₂H₃ClMg
MW: 86.801 g/mol
Boiling Pt: 66 °C
Density: 0.98 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00013594
CAS Number: 3536-96-7
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Clear to hazy yellow to dark brown Solution
Titration Active base 1.68 to 1.72 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR