Bismuth (III) oxyfluoride 97%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: fluorooxobismuthine, Bismuth (III) Fluoride Oxide, Fluorobismuth oxide

AC45382-0250 AC45382-1000
TS45382-0250EA 81 USD
TS45382-0250 TS45382-1000
Bismuth (III) oxyfluoride 97%, pure
Bismuth (III) oxyfluoride
Formula: BiFO
MW: 243.98 g/mol
Melting Pt: 500 °C
Storage Temperature: Ambient
MDL Number: MFCD00152288
CAS Number: 13520-72-4
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Appearance White to almost white Powder
Titration Complexometric 96.0 to 104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR