Lithium aluminium di-n-butylamide 0.16 M in 1,2-dimethoxyethane

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lithium tetrakis(N-butyl-1-butanaminato)aluminate(1-), Aluminate(1-), tetrakis(N-butyl-1-butanaminato)-, lithium (1:1)

AC44919-1000 AC44919-5000
TS44919-1000EA 92 USD
TS44919-1000 TS44919-5000
Lithium aluminium di-n-butylamide 0.16 M in 1,2-dimethoxyethane
Lithium aluminium di-n-butylamide
Formula: C₃₂H₇₂AlLiN₄
Melting Pt: –54 °C
Density: 0.86 g/cm³
Flash Pt: –6 °C
Storage Temperature: Ambient
MDL Number: MFCD00051918
CAS Number: 15405-86-4
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Solution (May develop some turbidity or precipitate)
Strength 0.14 to 0.18 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR