N-TIPS indole-3-boronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-1-triisopropylsilanyl-1H-indole, 2-[1-(Triisopropylsilyl)-1H-indole-3-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

AC44103-0010 AC44103-0050
TS44103-0010EA 186 USD
TS44103-0010 TS44103-0050
N-TIPS indole-3-boronic acid pinacol ester 97%
N-TIPS indole-3-boronic acid pinacol ester
Formula: C₂₃H₃₈BNO₂Si
MW: 399.457 g/mol
Melting Pt: 111…115 °C
Storage Temperature: Ambient
MDL Number: MFCD16659107
CAS Number: 476004-82-7

Order Now

Specification Test Results

Appearance White to off-white to pink Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR