(1,3-Dioxolan-2-ylmethyl)magnesium bromide 0.5 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Magnesium 2-methanidyl-1,3-dioxolane Bromide, magnesium(2+) ion 1,3-dioxolan-2-ylmethanide bromide

AC43886-1000 AC43886-8000
TS43886-1000EA 112 USD
TS43886-1000 TS43886-8000
(1,3-Dioxolan-2-ylmethyl)magnesium bromide 0.5 M in THF, AcroSeal™
(1,3-Dioxolan-2-ylmethyl)magnesium bromide
Formula: C₄H₇BrMgO₂
MW: 191.31 g/mol
Boiling Pt: 65…67 °C (760 mmHg)
Density: 0.94 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD01321271
CAS Number: 180675-22-3
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Light yellow to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 0.40 to 0.60 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR