Tetraammineplatinum(II) chloride hydrate 99.995% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: AKOS015924294, AC-24209

AC43700-0050 AC43700-5000
TS43700-0050EA 1008 USD
TS43700-0050 TS43700-5000
Tetraammineplatinum(II) chloride hydrate 99.995% (trace metals basis)
Tetraammineplatinum(II) chloride hydrate
Formula: Pt(NH₃)₄Cl₂·xH₂O
MW: 334.11 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149947
CAS Number: 108374-32-9

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Total trace metal impurities ≤50 ppm
Platinum (Pt) 51 to 59 % (typical)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR