1-Propenylmagnesium bromide 0.5 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (Z)-Prop-1-en-1-ylmagnesium bromide, (E)-Prop-1-en-1-ylmagnesium bromide

AC43467-1000 AC43467-8000
TS43467-1000EA 133 USD
TS43467-1000 TS43467-8000
1-Propenylmagnesium bromide 0.5 M in THF, AcroSeal™
1-Propenylmagnesium bromide
Formula: C₃H₅BrMg
Boiling Pt: 65 °C (760 mmHg)
Density: 0.95 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00209610
CAS Number: 14092-04-7
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Light yellow to orange to brown Solution
Titration Grignard 0.45 to 0.55 M
Melting point 138 to 142 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR