Dilithium tetrabromonickelate(II) 0.5 M in THF, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC43454-1000
AC43454-8000
TS43454-1000EA
47
USD
TS43454-1000
TS43454-8000
Dilithium tetrabromonickelate(II) 0.5 M in THF, AcroSeal™
Dilithium tetrabromonickelate(II)
Formula:
Br₄Li₂Ni Density: 1.065 g/cm³ Flash Pt: –18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192058 CAS Number: 13826-95-4 UN: 1993 ADR: 3,II |
Specification Test Results
Appearance | Blue-green to brown Solution |
Titration Argentometric | 0.45 to 0.55 M |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...