Cesium methanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methanesulphonic acid cesium salt

AC43158-0050 AC43158-0250
TS43158-0050EA 91 USD
TS43158-0050 TS43158-0250
Cesium methanesulfonate 98%
Cesium methanesulfonate
Formula: CH₃CsO₃S
MW: 227.997 g/mol
Melting Pt: 262…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00075523
CAS Number: 2550-61-0

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR