Sodium phosphite dibasic pentahydrate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Etidronate disodium Related Compound A, United States Pharmacopeia (USP) Reference Standard, METAPHOSPHORIC ACID SODIUM SALT
AC42850-0010
AC42850-2500
TS42850-0010EA
100
USD
TS42850-0010
TS42850-2500
Sodium phosphite dibasic pentahydrate 98%
Sodium phosphite dibasic pentahydrate
Formula:
H₁₁Na₂O₈P MW: 216.033 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150354 CAS Number: 13517-23-2 |
Specification Test Results
Appearance | Transparent white to white Crystals and or bars |
Titration with HCl | ≥97.5 % |
Chloride (Cl) | ≤50 ppm |
Sulfate (SO4) | ≤50 ppm |
Iron (Fe) | ≤10 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...