Magnesium phosphate tribasic hydrate 98+%, pure

Supplier: Thermo Scientific Chemicals

AC39194-0010 AC39194-2500
TS39194-0010EA 84 USD
TS39194-0010 TS39194-2500
Magnesium phosphate tribasic hydrate 98+%, pure
Magnesium phosphate tribasic hydrate
Formula: H₂Mg₃O₉P₂
MW: 280.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150251
CAS Number: 53408-95-0

Order Now

Specification Test Results

Appearance White Powder
Titration excess HCl 98.0 to 102.0 % (On ignited basis)
Loss on ignition ≤32.0 % (425°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR