Palladium, black 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC19515-0010
AC19515-0050
TS19515-0010EA
331
USD
TS19515-0010
TS19515-0050
Palladium, black 98%
Palladium
Formula:
Pd Boiling Pt: 3167 °C (1013 hPa) Melting Pt: 1555 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 UN: 3089 ADR: 4.1,II |
Specification Test Results
Appearance | Black Powder |
Assay | ≥97.5 % (Pd) |
Calcium (Ca) | ≤25 ppm |
Copper (Cu) | ≤25 ppm |
Gold (Au) | ≤50 ppm |
Iridium (Ir) | ≤50 ppm |
Iron (Fe) | ≤25 ppm |
Lead (Pb) | ≤25 ppm |
Magnesium (Mg) | ≤25 ppm |
Platinum (Pt) | ≤50 ppm |
Rhodium (Rh) | ≤50 ppm |
Ruthenium (Ru) | ≤50 ppm |
Silicon (Si) | ≤50 ppm |
Silver (Ag) | ≤50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...