Palladium 10% on activated carbon wetted with ca. 50% water, unreduced

Supplier: Thermo Scientific Chemicals

AC42298-0010 AC42298-0100 AC42298-0250 AC42298-1000
TS42298-0010EA 55 USD
TS42298-0010 TS42298-0100 TS42298-0250 TS42298-1000
Palladium 10% on activated carbon wetted with ca. 50% water, unreduced
Palladium
Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now

Specification Test Results

Appearance Black Powder
Assay 9.7 to 10.3 % (Pd)
Loss on drying 45 % to 55 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR