Palladium 10% on activated carbon, dry, unreduced
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC19503-0010
AC19503-0100
AC19503-0500
AC19503-2500
195030100
TS19503-0010EA
37
USD
TS19503-0010
TS19503-0100
TS19503-0500
TS19503-2500
TS195030100
Palladium 10% on activated carbon, dry, unreduced
Palladium
Formula:
Pd MW: 106.420 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 UN: 3178 ADR: 4.1,III |
Specification Test Results
Appearance | Black Powder |
Assay | 9.7 to 10.3 % (Pd) |
Loss on drying | ≤5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...