Palladium (5% Pd) on activated carbon, unreduced

Supplier: Thermo Scientific Chemicals
Warning

AC19502-0100 AC19502-1000 AC19502-5000
TS19502-0100EA 165 USD
TS19502-0100 TS19502-1000 TS19502-5000
Palladium (5% Pd) on activated carbon, unreduced
Palladium
Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 3178
ADR: 4.1,III

Order Now

Specification Test Results

Appearance Black Powder
Assay 4.8 to 5.2 % (Pd) On dry basis
Loss on drying ≤3 % (120°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR