Sodium metatungstate hydrate, crystalline

Supplier: Thermo Scientific Chemicals
Danger

AC30409-0010 AC30409-0500 AC30409-2500
TS30409-0010EA 1289 USD
TS30409-0010 TS30409-0500 TS30409-2500
Sodium metatungstate hydrate, crystalline
Sodium metatungstate hydrate
Formula: H₂Na₆O₄₀W₁₂·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00150663
CAS Number: 652154-12-6

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or beads
Assay ≥85 % (WO3)
Stabilizer ≤200 ppm (Cr-salt) Typical

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR