Tetracarbonyldi-µ-chlorodirhodium(I) 97%

Supplier: Thermo Scientific Chemicals
Warning

AC19760-0010 AC19760-2500
TS19760-0010EA 687 USD
TS19760-0010 TS19760-2500
Tetracarbonyldi-µ-chlorodirhodium(I) 97%
Tetracarbonyldi-µ-chlorodirhodium(I)
Formula: C₄Cl₂O₄Rh₂
Melting Pt: 121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00135610
CAS Number: 14523-22-9

Order Now

Specification Test Results

Appearance Red to brown Crystalline powder or crystals or needles
Infrared spectrum Conforms
Assay ≥96.0 % (ex Rh)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR