Potassium hexacyanocobaltate(III) 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Tripotassium hexacyanocobaltate(III)

AC20392-0050 AC20392-1000 AC20392-5000
TS20392-0050EA 23 USD
TS20392-0050 TS20392-1000 TS20392-5000
Potassium hexacyanocobaltate(III) 95%
Potassium hexacyanocobaltate(III)
Formula: C₆CoK₃N₆
MW: 332.332 g/mol
Density: 1.87 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011391
CAS Number: 13963-58-1

Order Now

Specification Test Results

Appearance Light yellow to light brown Crystalline solid
Assay ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR