Dicarbonyl(2,4-pentanedionato)rhodium(I)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (Acetylacetonato)dicarbonylrhodium(I), Dicarbonylacetylacetonato rhodium(I)

AC36395-0025 AC36395-5000
TS36395-0025EA 2320 USD
TS36395-0025 TS36395-5000
Dicarbonyl(2,4-pentanedionato)rhodium(I)
Dicarbonyl(2,4-pentanedionato)rhodium(I)
Formula: C₇H₈O₄Rh
MW: 259.04 g/mol
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00009884
CAS Number: 14874-82-9
UN: 3467
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Green-red Flakes or needles
Infrared spectrum Conforms
Rhodium (Rh) ≥39.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR