Cyclohexylmagnesium chloride 1.3 M in THF/toluene, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC37734-1000 AC37734-8000
TS37734-1000EA 59 USD
TS37734-1000 TS37734-8000
Cyclohexylmagnesium chloride 1.3 M in THF/toluene, AcroSeal™
Cyclohexylmagnesium chloride
Formula: C₆H₁₁ClMg
Density: 0.95 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00003816
CAS Number: 931-51-1
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear dark brown Solution (May develop some turbidity or precipitate)
Titration Grignard 1.26 to 1.40 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR