Ammonium bis(oxalato)oxotitanate(IV) hydrate 98%, pure

Supplier: Thermo Scientific Chemicals
Warning

AC21273-0100 AC21273-1000
TS21273-0100EA 93 USD
TS21273-0100 TS21273-1000
Ammonium bis(oxalato)oxotitanate(IV) hydrate 98%, pure
Ammonium bis(oxalato)oxotitanate(IV) hydrate
Formula: (NH₄)₂TiO(C₂O₄)₂·xH₂O (x≈1)
MW: 276.01 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149620
CAS Number: 10580-03-7

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Titration with KMnO4 ≥97.5 % (Calculated on oxalate)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR