(3-Bromopropoxy)-tert-butyldimethylsilane 97% stabilised

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-Bromopropoxy)(tert-butyl)dimethylsilane

Stabilization: Stabilised with sodium carbonate (Na₂CO₃)

AC43869-0050 AC43869-0250
TS43869-0050EA 118 USD
TS43869-0050 TS43869-0250
(3-Bromopropoxy)-tert-butyldimethylsilane 97% stabilised
(3-Bromopropoxy)-tert-butyldimethylsilane
Formula: C₉H₂₁BrOSi
Boiling Pt: 182 °C (760 mmHg)
Density: 1.093 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00216589
CAS Number: 89031-84-5

Order Now

Specification Test Results

Appearance Colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR