Poly(methylhydrosiloxane) trimethylsilyl terminated

Supplier: Thermo Scientific Chemicals

AC44010-0250 AC44010-1000
TS44010-0250EA 58 USD
TS44010-0250 TS44010-1000
Poly(methylhydrosiloxane) trimethylsilyl terminated
Poly(methylhydrosiloxane)
Formula: (CH₃)₃SiO[(CH₃)HSiO]nSi(CH₃)
Boiling Pt: ∼140 °C (0.003 hPa)
Melting Pt: –55 °C
Density: 1.006 g/cm³
Flash Pt: 204 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00084478
CAS Number: 63148-57-2

Order Now

Specification Test Results

Appearance Colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3960 to 1.4000 (20°C, 589 nm)
Viscosity 15 to 25 cSt

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR