Zinc oxide ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Zinci oxidum, Zinc (II) oxide
AC42274-0010
AC42274-0051
AC42274-1000
AC42274-5000
TS42274-0010EA
73
USD
TS42274-0010
TS42274-0051
TS42274-1000
TS42274-5000
Zinc oxide ACS
Zinc oxide
Formula:
ZnO MW: 81.39 g/mol Melting Pt: 1975 °C Density: 5.6 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011300 CAS Number: 1314-13-2 EINECS: 215-222-5 UN: 3077 ADR: 9 III REACH: 01-2119463881-32 Merck Index: 13,10200 |
Specification Test Results
Appearance | White to off-white Powder |
Titration Complexometric | ≥99.0 % |
Alkalinity | passes test |
Chloride (Cl) | ≤10 ppm |
Nitrate (NO3) | ≤30 ppm |
Calcium (Ca) | ≤50 ppm |
Iron (Fe) | ≤10 ppm |
Lead (Pb) | ≤50 ppm |
Magnesium (Mg) | ≤50 ppm |
Manganese (Mn) | ≤5 ppm |
Potassium (K) | ≤100 ppm |
Sodium (Na) | ≤500 ppm |
Sulfur compounds (as SO4) | ≤0.01 % |
Insoluble matter | ≤0.01 % (in dilute H2SO4) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...