Barium chloranilate trihydrate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chloranilic acid barium salt, Barium 3,6-Dichloro-1,4-dioxo-3,6-cyclohexadiene-2,5-diolate

AC40166-0250 AC40166-1000
TS40166-0250EA 213 USD
TS40166-0250 TS40166-1000
Barium chloranilate trihydrate
Barium chloranilate trihydrate
Formula: C₆H₂Cl₂O₄
MW: 344.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150175
CAS Number: 32458-20-1
UN: 1564
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Brown Powder
Infrared spectrum Conforms
Suitability (for Sulfate determination) Passes test
Titration with NaOH 94 to 108 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR