(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1R,2R)-(-)-1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)cobalt (II), (R,R)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II), (R,R)-Jacobsen's catalyst cobalt(II) complex

AC32995-0010 AC32995-0050 AC32995-0250
TS32995-0010EA 41 USD
TS32995-0010 TS32995-0050 TS32995-0250
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II) 98%
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II)
Formula: C₃₆H₅₂CoN₂O₂
MW: 603.74 g/mol
Melting Pt: >350 °C
Storage Temperature: Ambient
MDL Number: MFCD01631277
CAS Number: 176763-62-5

Order Now

Specification Test Results

Appearance Orange to red-brown Powder
Infrared spectrum Conforms
Elemental Analysis ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR