Indium(III) acetate, anhydrous 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

AC43698-0100 AC43698-0500
TS43698-0100EA 121 USD
TS43698-0100 TS43698-0500
Indium(III) acetate, anhydrous 99.99% (trace metals basis)
Indium(III) acetate
Formula: C₂H₄InO₂
MW: 174.87 g/mol
Melting Pt: 270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00054347
CAS Number: 25114-58-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Complexometric 35.3 to 43.3 % (In)
Total trace metal impurities ≤100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR