Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(A-taPhos)2PdCl2, PdCl2(Amphos)2, Bis(di-tert-butyl(4-dimethylaminophenyl)phosphine)dichloropalladium(II), Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium, Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II)
44789-0050
AC44789-0010
TS44789-0050EA
688
USD
TS44789-0050
TS44789-0010
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) 95%
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II)
Formula:
C₃₂H₅₆Cl₂N₂P₂Pd Storage Temperature: Ambient |
MDL Number:
MFCD09265123 CAS Number: 887919-35-9 |
Specification Test Results
Appearance | Yellow to dark yellow to dark orange Powder and/or chunks |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...