Isopropenylmagnesium bromide solution 0.5 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC42775-1000 AC42775-8000
TS42775-1000EA 96 USD
TS42775-1000 TS42775-8000
Isopropenylmagnesium bromide solution 0.5 M in THF, AcroSeal™
Isopropenylmagnesium bromide solution
Formula: C₃H₅BrMg
Boiling Pt: 67 °C (760 mmHg)
Density: 0.927 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
CAS Number: 13291-18-4
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Yellow to gray to brown Solution
Titration Active base 0.45 to 0.60 M
Melting point 37 to 41 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR