3-Butenylmagnesium bromide 0.5 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC42746-1000 AC42746-8000
TS42746-1000EA 347 USD
TS42746-1000 TS42746-8000
3-Butenylmagnesium bromide 0.5 M in THF, AcroSeal™
3-Butenylmagnesium bromide
Formula: C₄H₇BrMg
Boiling Pt: 65 °C (760 mmHg)
Density: 0.937 g/cm³
Flash Pt: –30 °C
Storage Temperature: Ambient
CAS Number: 7103-09-5
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Gray to brown Solution
Titration Grignard 0.40 to 0.60 M
Identification Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR