4-(tert-Butyldimethylsilyloxy)cyclohexanone 97%

Supplier: Thermo Scientific Chemicals
Warning

AC42685-0010 AC42685-0050
TS42685-0010EA 38 USD
TS42685-0010 TS42685-0050
4-(tert-Butyldimethylsilyloxy)cyclohexanone 97%
4-(tert-Butyldimethylsilyloxy)cyclohexanone
Formula: C₁₂H₂₄O₂Si
Boiling Pt: 85 °C (1 mmHg)
Density: 0.909 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD06411307
CAS Number: 55145-45-4

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR