Europium(III) thenoyltrifluoroacetonate trihydrate 95%

Supplier: Thermo Scientific Chemicals

AC42319-0010 AC42319-0050
TS42319-0010EA 211 USD
TS42319-0010 TS42319-0050
Europium(III) thenoyltrifluoroacetonate trihydrate 95%
Europium(III) thenoyltrifluoroacetonate trihydrate
Formula: C₂₄H₂₁EuF₉O₉S₃
Melting Pt: 142…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00150912
CAS Number: 21392-96-1

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 142 to 147 °C
Elemental Analysis ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR