Potassium methyl sulfate 99%

Supplier: Thermo Scientific Chemicals
Danger

AC41489-0100 AC41489-1000
TS41489-0100EA 95 USD
TS41489-0100 TS41489-1000
Potassium methyl sulfate 99%
Potassium methyl sulfate
Formula: CH₃KO₄S
Melting Pt: 212…217 °C
Storage Temperature: Ambient
CAS Number: 562-54-9
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder or flakes
Infrared spectrum Conforms
Titration after Ion exchange ≥98.5 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR