(tert-Butyldimethylsiloxy)acetaldehyde 90%
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC39751-0010
AC39751-0050
TS39751-0010EA
87
USD
TS39751-0010
TS39751-0050
(tert-Butyldimethylsiloxy)acetaldehyde 90%
(tert-Butyldimethylsiloxy)acetaldehyde
Formula:
C₈H₁₈O₂Si Boiling Pt: 156…157 °C (760 mmHg) Density: 0.915 g/cm³ Flash Pt: 60 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01321229 CAS Number: 102191-92-4 UN: 1989 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to straw yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥88 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...