Chromium(III) chloride tetrahydrofuran complex (1:3) 98%

Supplier: Thermo Scientific Chemicals
Warning

AC38985-0010 AC38985-0100
TS38985-0010EA 36 USD
TS38985-0010 TS38985-0100
Chromium(III) chloride tetrahydrofuran complex (1:3) 98%
Chromium(III) chloride tetrahydrofuran complex (1:3)
Formula: C₁₂H₂₄Cl₃CrO₃
MW: 374.667 g/mol
Melting Pt: 143 °C
Storage Temperature: Ambient
CAS Number: 10170-68-0
UN: 1325
ADR: 4.1 II

Order Now

Specification Test Results

Appearance Purple Crystalline powder
Infrared spectrum Conforms
Titration with Na2S2O3 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR