(1-Ethoxycyclopropoxy)trimethylsilane 98%

Supplier: Thermo Scientific Chemicals
Warning

AC36890-0050 AC36890-0250
TS36890-0050EA 92 USD
TS36890-0050 TS36890-0250
(1-Ethoxycyclopropoxy)trimethylsilane 98%
(1-Ethoxycyclopropoxy)trimethylsilane
Formula: C₈H₁₈O₂Si
MW: 174.315 g/mol
Boiling Pt: 50…53 °C (22 mmHg)
Density: 0.867 g/cm³
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00074986
CAS Number: 27374-25-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4090 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR